ENAH38A2CE73 Enamine

2-(3-{[(tert-butoxy)carbonyl]amino}adamantan-1-yl)acetic acid

  • Cas Number : 2309448-62-0
  • Molecular Weight : 309.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6754332

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6754332
Availability details for
Quantity
  • Availability
  • Add to Cart