ENAH38A2CE76 Enamine

methyl 2-[(piperidin-4-yl)methoxy]acetate hydrochloride

  • MDL number MFCD31810810
  • Cas Number : 2305254-75-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6754340

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6754340
Availability details for
Quantity
  • Availability
  • Add to Cart