ENAH38A2CE7A Enamine

(2R)-1-(2,3-dihydro-1H-inden-5-yl)propan-2-amine hydrochloride

  • MDL number MFCD31811085
  • Cas Number : 2305184-97-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6754449

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6754449
Availability details for
Quantity
  • Availability
  • Add to Cart