ENAH38A2CE7E Enamine

(dimethylphosphoryl)benzene

  • MDL number MFCD00045692
  • Cas Number : 10311-08-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6756635

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6756635
Availability details for
Quantity
  • Availability
  • Add to Cart