ENAH38A2CE7F Enamine

(5r,8r)-3-methyl-2,4-dioxo-1,3-diazaspiro[4.5]decane-8-carboxylic acid

  • MDL number MFCD31810814
  • Cas Number : 2305147-99-1
  • Molecular Weight : 226.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6756746

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-6756746
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart