ENAH38A2CE82 Enamine

4-(hydroxymethyl)-4-methyloxolan-2-one

  • MDL number MFCD30726591
  • Cas Number : 1142950-59-1
  • Molecular Weight : 130.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6756995

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6756995
Availability details for
Quantity
  • Availability
  • Add to Cart