ENAH38A2CE85 Enamine

9-benzyl-1-oxa-3,9-diazaspiro[5.5]undecan-2-one

  • MDL number MFCD28040600
  • Cas Number : 54981-06-5
  • Molecular Weight : 260.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6757605

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6757605
Availability details for
Quantity
  • Availability
  • Add to Cart