ENAH38A2CE86 Enamine

6-ethyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carbonitrile

  • MDL number MFCD01055008
  • Cas Number : 5915-17-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6757607

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6757607
Availability details for
Quantity
  • Availability
  • Add to Cart