ENAH38A2CE89 Enamine

N'-hydroxy-4-(trifluoromethyl)thiophene-3-carboximidamide

  • Cas Number : 2309485-09-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6757615

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6757615
Availability details for
Quantity
  • Availability
  • Add to Cart