ENAH38A2CE8B Enamine

1-cyclopentyl-1,2,3,4-tetrahydropyrimidine-2,4-dione

  • MDL number MFCD28364736
  • Cas Number : 13345-72-7
  • Molecular Weight : 180.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6757617

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6757617
Availability details for
Quantity
  • Availability
  • Add to Cart