ENAH38A2CE8E Enamine

1-(2,2-dimethoxyethyl)-2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carbonitrile

  • MDL number MFCD30005268
  • Cas Number : 2098101-39-2
  • Molecular Weight : 225.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6757623

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6757623
Availability details for
Quantity
  • Availability
  • Add to Cart