ENAH38A2CE90 Enamine

2-(1-benzylpiperidin-4-yl)-2-phenylacetonitrile

  • MDL number MFCD00023760
  • Cas Number : 7254-21-9
  • Molecular Weight : 290.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6757625

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6757625
Availability details for
Quantity
  • Availability
  • Add to Cart