ENAH38A2CE91 Enamine

2-(1-benzylpiperidin-4-ylidene)-2-phenylacetonitrile

  • MDL number MFCD02728495
  • Cas Number : 6517-69-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6757626

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6757626
Availability details for
Quantity
  • Availability
  • Add to Cart