ENAH38A2CE99 Enamine

methyl 5-[(dimethylamino)methyl]-1,2-oxazole-3-carboxylate

  • MDL number MFCD10485376
  • Cas Number : 932848-17-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6758598

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6758598
Availability details for
Quantity
  • Availability
  • Add to Cart