ENAH38A2CEA0 Enamine

tert-butyl N-(3-carbamoylazetidin-3-yl)-N-methylcarbamate hydrochloride

  • MDL number MFCD31811092
  • Cas Number : 2305255-46-1
  • Molecular Weight : 265.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6758614

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6758614
Availability details for
Quantity
  • Availability
  • Add to Cart