ENAH38A2CEA3 Enamine

2-bromo-4-[(tert-butoxy)carbonyl]benzoic acid

  • MDL number MFCD30188715
  • Cas Number : 2089325-57-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6758620

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6758620
Availability details for
Quantity
  • Availability
  • Add to Cart