ENAH38A2CEA6 Enamine

4-{[(4,6-dimethylpyrimidin-2-yl)sulfanyl]methyl}-N'-[1-(4-methyl-3-nitrophenyl)ethylidene]benzohydrazide

  • Cas Number : 1013528-93-2
  • Molecular Weight : 449.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 205-206
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6758638

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-6758638
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart