ENAH38A2CEA7 Enamine

2-(pyridin-2-yl)-2H-1,2,3-triazol-4-amine

  • MDL number MFCD26128528
  • Cas Number : 1710344-48-1
  • Molecular Weight : 161.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6758639

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6758639
Availability details for
Quantity
  • Availability
  • Add to Cart