ENAH38A2CEB1 Enamine

dimethyl[(trimethylsilyl)imino]-lambda6-sulfanone

  • MDL number MFCD31811096
  • Cas Number : 24989-35-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6758722

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6758722
Availability details for
Quantity
  • Availability
  • Add to Cart