ENAH38A2CEBC Enamine

1-oxo-1,2-dihydrophthalazine-6-carboxylic acid

  • MDL number MFCD28679116
  • Cas Number : 102126-65-8
  • Molecular Weight : 190.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6758811

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6758811
Availability details for
Quantity
  • Availability
  • Add to Cart