ENAH38A2CEC3 Enamine

1-methyl-4-(trifluoromethyl)-1H-imidazol-2-amine

  • MDL number MFCD31915782
  • Cas Number : 2109509-60-4
  • Molecular Weight : 165.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6758872

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6758872
Availability details for
Quantity
  • Availability
  • Add to Cart