ENAH38A2CED0 Enamine

2-bromo-4-chloro-1-ethyl-1H-imidazole-5-carbonitrile

  • MDL number MFCD31811115
  • Cas Number : 2091537-29-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6758934

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6758934
Availability details for
Quantity
  • Availability
  • Add to Cart