ENAH38A2CEDC Enamine

1-[4-(trifluoromethyl)thiophen-2-yl]ethan-1-one

  • MDL number MFCD22060965
  • Cas Number : 1368164-70-8
  • Molecular Weight : 194.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6759012

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6759012
Availability details for
Quantity
  • Availability
  • Add to Cart