ENAH38A2CEDD Enamine

methyl 2-[3-(chlorosulfonyl)cyclopentyl]acetate

  • MDL number MFCD31810886
  • Cas Number : 2168775-82-2
  • Molecular Weight : 240.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form oil
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6759014

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6759014
Availability details for
Quantity
  • Availability
  • Add to Cart