ENAH38A2CEDE Enamine

2-butyl-2H-1,2,3-triazole

  • MDL number MFCD31811122
  • Cas Number : 1799613-65-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6759028

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6759028
Availability details for
Quantity
  • Availability
  • Add to Cart