ENAH38A2CEE3 Enamine

tert-butyl 4-(4-hydroxy-5-oxopyrrolidin-2-yl)piperidine-1-carboxylate

  • MDL number MFCD31915350
  • Cas Number : 2309468-12-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6759059

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6759059
Availability details for
Quantity
  • Availability
  • Add to Cart