ENAH38A2CEE6 Enamine

3-(2-aminoacetamido)benzamide hydrochloride

  • MDL number MFCD31810888
  • Cas Number : 2305256-04-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6759072

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6759072
Availability details for
Quantity
  • Availability
  • Add to Cart