ENAH38A2CEF4 Enamine

4-{[methyl(phenyl)amino]methyl}benzoic acid

  • MDL number MFCD09046936
  • Cas Number : 102244-39-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6759208

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6759208
Availability details for
Quantity
  • Availability
  • Add to Cart