ENAH38A2CEF9 Enamine

5-bromo-2-ethyl-2H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD31810933
  • Cas Number : 1372711-57-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6759216

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6759216
Availability details for
Quantity
  • Availability
  • Add to Cart