ENAH38A2CEFF Enamine

3-benzylpentanedioic acid

  • MDL number MFCD02066184
  • Cas Number : 32386-49-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6759246

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6759246
Availability details for
Quantity
  • Availability
  • Add to Cart