ENAH38A2CF07 Enamine

(1E)-1-bromocyclooct-1-ene

  • MDL number MFCD28063955
  • Cas Number : 61815-42-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6759278

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6759278
Availability details for
Quantity
  • Availability
  • Add to Cart