ENAH38A2CF08 Enamine

1-(5-fluoro-1H-indol-3-yl)-2-methylpropan-2-amine

  • MDL number MFCD18800910
  • Cas Number : 118096-08-5
  • Molecular Weight : 206.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6497635

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6497635
Availability details for
Quantity
  • Availability
  • Add to Cart