ENAH38A2CF10 Enamine

1-benzyl-5-chloro-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD01109285
  • Cas Number : 15212-80-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6759330

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6759330
Availability details for
Quantity
  • Availability
  • Add to Cart