ENAH38A2CF31 Enamine

1,5,5-trimethylpyrrolidin-3-amine dihydrochloride

  • MDL number MFCD31810996
  • Cas Number : 2305252-71-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6759582

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6759582
Availability details for
Quantity
  • Availability
  • Add to Cart