ENAH38A2CF36 Enamine

4-(2-aminoacetamido)benzamide dihydrochloride

  • MDL number MFCD31811136
  • Cas Number : 2305255-41-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6759597

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6759597
Availability details for
Quantity
  • Availability
  • Add to Cart