ENAH38A2CF3B Enamine

{2-oxabicyclo[2.1.1]hexan-4-yl}methanol

  • MDL number MFCD28650665
  • Cas Number : 1782594-07-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6759622

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6759622
Availability details for
Quantity
  • Availability
  • Add to Cart