ENAH38A2CF4D Enamine

[(2S)-1-(carboxy)pyrrolidin-2-yl]methyl acetate

  • MDL number MFCD28006029
  • Cas Number : 1296132-58-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6759764

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6759764
Availability details for
Quantity
  • Availability
  • Add to Cart