ENAH38A2CF4F Enamine

N,N-bis(propan-2-yl)-1,3,2-dithiaphospholan-2-amine

  • MDL number MFCD25959103
  • Cas Number : 145498-60-8
  • Molecular Weight : 223.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140-141
Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6759772

Safety Information

signalword Danger
pcodes P260,P261,P264,P270,P271,P280,P301+P310,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P330,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-6759772
pictograms GHS05,GHS06
hstatements H301,H314,H335
Availability details for
Quantity
  • Availability
  • Add to Cart