ENAH38A2CF5D Enamine

7-oxo-5-phenyl-4H,7H-pyrazolo[1,5-a]pyrimidine-2-carboxylic acid

  • MDL number MFCD31915419
  • Cas Number : 632365-48-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6759827

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6759827
Availability details for
Quantity
  • Availability
  • Add to Cart