ENAH38A2CF5E Enamine

5-{[(9H-fluoren-9-yl)methoxy]carbonyl}-8-oxa-5-azaspiro[3.5]nonane-7-carboxylic acid

  • MDL number MFCD31915420
  • Cas Number : 2309448-55-1
  • Molecular Weight : 393.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6759830

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6759830
Availability details for
Quantity
  • Availability
  • Add to Cart