ENAH38A2CF64 Enamine

1-[(tert-butoxy)carbonyl]-8-thia-1-azaspiro[4.5]decane-4-carboxylic acid

  • MDL number MFCD31811158
  • Cas Number : 2305254-87-7
  • Molecular Weight : 301.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6759856

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6759856
Availability details for
Quantity
  • Availability
  • Add to Cart