ENAH38A2CF68 Enamine

N-cyclobutylbenzamide

  • MDL number MFCD20360625
  • Cas Number : 120218-51-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6759864

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6759864
Availability details for
Quantity
  • Availability
  • Add to Cart