ENAH38A2CF6E Enamine

sodium 4-(difluoromethyl)-5-(ethoxycarbonyl)pyrimidin-2-olate

  • MDL number MFCD31811160
  • Cas Number : 2305251-58-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6759982

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6759982
Availability details for
Quantity
  • Availability
  • Add to Cart