ENAH38A2CF8B Enamine

6,6-dimethylpiperidin-3-ol hydrochloride

  • MDL number MFCD31811066
  • Cas Number : 2305253-59-0
  • Molecular Weight : 165.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6760206

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6760206
Availability details for
Quantity
  • Availability
  • Add to Cart