ENAH38A2CF8D Enamine

6-(trifluoromethoxy)-2,3,4,9-tetrahydro-1H-carbazol-1-one

  • MDL number MFCD31915790
  • Cas Number : 893410-19-0
  • Molecular Weight : 269.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6760210

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6760210
Availability details for
Quantity
  • Availability
  • Add to Cart