ENAH38A2CF94 Enamine

2-[(tert-butoxy)carbonyl]-8-thia-2-azaspiro[4.5]decane-3-carboxylic acid

  • MDL number MFCD31811072
  • Cas Number : 2305254-61-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6760238

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6760238
Availability details for
Quantity
  • Availability
  • Add to Cart