ENAH38A2CF9C Enamine

(3aR,6aS)-octahydropentalen-2-one

  • MDL number MFCD11878434
  • Cas Number : 19915-11-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6760303

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6760303
Availability details for
Quantity
  • Availability
  • Add to Cart