ENAH38A2CFA0 Enamine

(1S)-1-(1H-1,2,4-triazol-3-yl)ethan-1-amine dihydrochloride

  • Cas Number : 1150263-83-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6760326

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6760326
Availability details for
Quantity
  • Availability
  • Add to Cart