ENAH38A2CFA2 Enamine

6-chloro-2-(methylsulfanyl)-[1,3]thiazolo[4,5-b]pyridine

  • MDL number MFCD27995291
  • Cas Number : 488742-51-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6760332

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6760332
Availability details for
Quantity
  • Availability
  • Add to Cart