ENAH38A2CFAD Enamine

4-methyl-1,2-diazetidin-3-one; 4-methylbenzene-1-sulfonic acid

  • MDL number MFCD31810788
  • Cas Number : 79289-51-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data TsOH
Storage temp. -10
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6762240

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6762240
Availability details for
Quantity
  • Availability
  • Add to Cart